Anti-DAAM1 Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A45371-100UL A45371-50UL
77709-325EA 334.95 USD
77709-325 77709-326
Anti-DAAM1 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:2000, WB:1:200-1:1000, IHC:1:50-1:200

Type:
Antigen: DAAM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR