Anti-APX1 Rabbit Polyclonal Antibody (FITC)

Supplier: AFG BIOSCIENCE LLC
A45336-50UG A45336-100UG
77709-261EA 228.38 USD
77709-261 77709-260
Anti-APX1 Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: APX1
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Arabidopsis thaliana
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR