Anti-TPM2 Rabbit Polyclonal Antibody (FITC)

Supplier: AFG BIOSCIENCE LLC
A45310-50UG A45310-100UG
77709-216EA 228.38 USD
77709-216 77709-215
Anti-TPM2 Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: TPM2
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR