Anti-PIK3CG Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A45280-50UG A45280-100UG
77709-160EA 228.38 USD
77709-160 77709-159
Anti-PIK3CG Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IF:1:50-1:200

Type:
Antigen: PIK3CG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR