Anti-FN1 Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A45231-100UG A45231-50UG
77709-071EA 334.95 USD
77709-071 77709-072
Anti-FN1 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: FN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR