Anti-TRIM3 Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A45196-50UL A45196-100UL
77709-010EA 228.38 USD
77709-010 77709-009
Anti-TRIM3 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: TRIM3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR