Anti-STAT4 Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A45176-50UL A45176-100UL
77708-971EA 228.38 USD
77708-971 77708-970
Anti-STAT4 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:3000, IHC:1:50-1:200

Type:
Antigen: STAT4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR