Anti-CRKL Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A45144-50UL A45144-100UL
77708-909EA 228.38 USD
77708-909 77708-908
Anti-CRKL Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, WB:1:500-1:2000, IHC:1:25-1:100

Type:
Antigen: CRKL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR