Anti-argF Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A45123-50UG A45123-100UG
77708-876EA 228.38 USD
77708-876 77708-875
Anti-argF Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: argF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Escherichia coli
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR