Anti-HIF1A Rabbit Polyclonal Antibody (Biotinylated)

Supplier: AFG BIOSCIENCE LLC
A45094-100UG A45094-50UG
77708-827EA 334.95 USD
77708-827 77708-828
Anti-HIF1A Rabbit Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: HIF1A
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR