Anti-GPC3 Rabbit Polyclonal Antibody (HRP)

Supplier: AFG BIOSCIENCE LLC
A45064-50UL A45064-100UL
77708-773EA 228.38 USD
77708-773 77708-772
Anti-GPC3 Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: GPC3
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR