Anti-PLXNA4 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A48161-100UL A48161-50UL
77714-179EA 334.95 USD
77714-179 77714-180
Anti-PLXNA4 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: PLXNA4
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR