Anti-SULT1B1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A47721-50UL A47721-100UL
77713-429EA 228.38 USD
77713-429 77713-428
Anti-SULT1B1 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:10000, WB:1:1000-1:5000, IHC:1:25-1:100

Type:
Antigen: SULT1B1
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR