Anti-CBS Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A47575-100UL A47575-50UL
77713-182EA 334.95 USD
77713-182 77713-183
Anti-CBS Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, WB:1:200-1:1000, IHC:1:25-1:100

Type:
Antigen: CBS
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR