Anti-TEP1 Rabbit Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A46957-50UG A46957-100UG
77712-121EA 228.38 USD
77712-121 77712-120
Anti-TEP1 Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: TEP1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR