Anti-TEP1 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A46959-50UG A46959-100UG
77712-125EA 228.38 USD
77712-125 77712-124
Anti-TEP1 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: TEP1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR