Anti-SNAP23 Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A46292-50UL A46292-100UL
77710-936EA 228.38 USD
77710-936 77710-935
Anti-SNAP23 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: SNAP23
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR