Anti-SLC26A4 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A47289-100UL A47289-50UL
77712-698EA 334.95 USD
77712-698 77712-699
Anti-SLC26A4 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: SLC26A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR