Anti-PRKCA Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A46181-50UG A46181-100UG
77710-738EA 228.38 USD
77710-738 77710-737
Anti-PRKCA Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: PRKCA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR