Anti-NEU1 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG BIOSCIENCE LLC
A46655-100UG A46655-50UG
77711-587EA 334.95 USD
77711-587 77711-588
Anti-NEU1 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: NEU1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR