Anti-MASP2 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A46869-100UL A46869-50UL
77711-965EA 334.95 USD
77711-965 77711-966
Anti-MASP2 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, IHC:1:25-1:100

Type:
Antigen: MASP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR