Anti-GDF1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A46239-100UL A46239-50UL
77710-843EA 334.95 USD
77710-843 77710-844
Anti-GDF1 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:2000, WB:1:200-1:1000

Type:
Antigen: GDF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR