Anti-CHEK2 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A47174-100UL A47174-50UL
77712-501EA 334.95 USD
77712-501 77712-502
Anti-CHEK2 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:2000, WB:1:200-1:1000

Type:
Antigen: CHEK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR