Anti-SCT Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A60376-50UL A60376-100UL
77716-412EA 228.38 USD
77716-412 77716-411
Anti-SCT Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, IHC:1:50-1:200

Type:
Antigen: SCT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR