Anti-APBA2 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A60062-100UL A60062-50UL
77715-890EA 334.95 USD
77715-890 77715-891
Anti-APBA2 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, IHC:1:100-1:300

Type:
Antigen: APBA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR