Anti-GATA5 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A48804-100UL A48804-50UL
77715-309EA 334.95 USD
77715-309 77715-310
Anti-GATA5 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, IHC:1:50-1:200

Type:
Antigen: GATA5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR