Anti-met Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A48752-100UG A48752-50UG
77689-287EA 334.95 USD
77689-287 77715-219
Anti-met Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:500-1:5000

Type:
Antigen: met
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR