Anti-fas Rabbit Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A48647-100UL A48647-50UL
77715-038EA 334.95 USD
77715-038 77715-039
Anti-fas Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: fas
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR