Anti-IL4I1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A64969-100UG A64969-50UG
77690-427EA 334.95 USD
77690-427 77725-028
Anti-IL4I1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:2000-1:10000, IHC:1:20-1:200

Type:
Antigen: IL4I1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR