Anti-C12orf43 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A64821-50UG A64821-100UG
77724-748EA 228.38 USD
77724-748 77724-747
Anti-C12orf43 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:20-1:200

Type:
Antigen: C12orf43
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR