Anti-SYNCRIP Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A44981-100UL A44981-50UL
77708-620EA 334.95 USD
77708-620 77708-621
Anti-SYNCRIP Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, IHC:1:25-1:100

Type:
Antigen: SYNCRIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR