Anti-KLK15 Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A44976-50UL A44976-100UL
77708-613EA 228.38 USD
77708-613 77708-612
Anti-KLK15 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:5000, WB:1:200-1:1000

Type:
Antigen: KLK15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR