Anti-TG Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A44933-100UG A44933-50UG
77708-533EA 334.95 USD
77708-533 77708-534
Anti-TG Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: TG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR