Anti-TPO Rabbit Polyclonal Antibody (Biotinylated)

Supplier: AFG BIOSCIENCE LLC
A44908-50UG A44908-100UG
77708-490EA 228.38 USD
77708-490 77708-489
Anti-TPO Rabbit Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: TPO
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR