Anti-IAPP Rabbit Polyclonal Antibody (HRP)

Supplier: AFG BIOSCIENCE LLC
A44891-100UG A44891-50UG
77708-458EA 334.95 USD
77708-458 77708-459
Anti-IAPP Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: IAPP
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR