Anti-ROBO4 Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A44865-50UL A44865-100UL
77708-412EA 228.38 USD
77708-412 77708-411
Anti-ROBO4 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: ROBO4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR