Anti-ICA1 Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A44842-50UG A44842-100UG
77708-367EA 228.38 USD
77708-367 77708-366
Anti-ICA1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200

Type:
Antigen: ICA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR