Anti-IFNL3 Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A44795-50UL A44795-100UL
77708-282EA 228.38 USD
77708-282 77708-281
Anti-IFNL3 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:2000, WB:1:200-1:1000

Type:
Antigen: IFNL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR