Anti-TFF3 Rabbit Polyclonal Antibody (FITC)

Supplier: AFG BIOSCIENCE LLC
A44782-100UG A44782-50UG
77708-256EA 334.95 USD
77708-256 77708-257
Anti-TFF3 Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: TFF3
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR