Anti-CDX2 Rabbit Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A44679-100UG A44679-50UG
77708-077EA 334.95 USD
77708-077 77708-078
Anti-CDX2 Rabbit Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: CDX2
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR