Anti-NAPSA Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A44596-100UG A44596-50UG
77707-932EA 334.95 USD
77707-932 77707-933
Anti-NAPSA Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200

Type:
Antigen: NAPSA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR