Anti-LAMC1 Rabbit Polyclonal Antibody (HRP)

Supplier: AFG BIOSCIENCE LLC
A44567-100UG A44567-50UG
77707-878EA 334.95 USD
77707-878 77707-879
Anti-LAMC1 Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: LAMC1
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR