Anti-IL2RB Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A44531-100UL A44531-50UL
77707-814EA 334.95 USD
77707-814 77707-815
Anti-IL2RB Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, WB:1:500-1:2000, IHC:1:50-1:200

Type:
Antigen: IL2RB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR