Anti-NEFH Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A44377-50UL A44377-100UL
77707-552EA 228.38 USD
77707-552 77707-551
Anti-NEFH Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, IHC:1:50-1:200

Type:
Antigen: NEFH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR