Anti-HSPA12B Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A44334-100UG A44334-50UG
77707-472EA 334.95 USD
77707-472 77707-473
Anti-HSPA12B Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:20-1:200

Type:
Antigen: HSPA12B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR