Anti-RICTOR Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A44278-100UL A44278-50UL
77707-376EA 334.95 USD
77707-376 77707-377
Anti-RICTOR Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: RICTOR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR