Anti-TDGF1 Rabbit Polyclonal Antibody (Biotinylated)

Supplier: AFG BIOSCIENCE LLC
A44239-100UG A44239-50UG
77707-310EA 334.95 USD
77707-310 77707-311
Anti-TDGF1 Rabbit Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: TDGF1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR