Anti-S100B Rabbit Polyclonal Antibody (HRP)

Supplier: AFG BIOSCIENCE LLC
A44195-50UG A44195-100UG
77707-229EA 228.38 USD
77707-229 77707-228
Anti-S100B Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: S100B
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR