Anti-CD3E Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A44093-50UG A44093-100UG
77707-060EA 228.38 USD
77707-060 77707-059
Anti-CD3E Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200, IF:1:50-1:200

Type:
Antigen: CD3E
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR