Anti-CUL7 Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A44014-100UL A44014-50UL
77706-915EA 334.95 USD
77706-915 77706-916
Anti-CUL7 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, IHC:1:50-1:200

Type:
Antigen: CUL7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR